Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Formaldehyd dimethyl acetal ≥98%

Glass bottle, 2.500 mL
Katalog # A12055.0F
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
Formaldehyd dimethyl acetal ≥98%
Katalog # A12055.0F
CAS nummer:  109-87-5
Formaldehyd dimethyl acetal ≥98%
Formaldehyd dimethyl acetal ≥98%

Glass bottle, 2.500 mL

CAS nummer:  109-87-5
Price
1.210,00 kr
Log ind for tilgængelighed
1 * 2.500 mL
Product Details & Documents

Purity: 98% CAS No.: 109-87-5 Molecular Formula: C3H8O2 Formula Weight: 76.10 Flash Point: -17°(1°F) Refractive Index: 1.3545 Melting Point: -105° Boiling Point: 41-42° Beilstein Reference: 1697025 Merck Reference: 14,6012 UN No.: 1234 MDL No.: MFCD00008495

Synonymer:  Formal, Methylal, Dimethoxymethane
  • CAS nummer:109-87-5
  • Formel:CH₂(OCH₃)₂
  • MDL nummer:MFCD00008495
  • Molecular Weight:76.1 g/mol
  • Merck Index:14,6012
  • Kogende Pt:41…42 °C
  • Smeltende Pt:–105 °C
  • Tæthed:0,863
  • Flash Pt:–17 °C (1 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-714-2
  • FN:1234
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2.500 mL

Specifikationer