Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Guanin ≥98%

Glass bottle for solids, 100 g
Katalog # A12024.22
Irriterende
Leverandør:   Thermo Fisher Scientific
Guanin ≥98%
Katalog # A12024.22
CAS nummer:  73-40-5
Guanin ≥98%
Guanin ≥98%

Glass bottle for solids, 100 g

CAS nummer:  73-40-5
Price
1.600,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 73-40-5 Molecular Formula: C5H5N5O Formula Weight: 151.13 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 9680 Merck Reference: 14,4564 MDL No.: MFCD00071533

Synonymer:  2-Amino-1H-purin-6(7H)-one
  • CAS nummer:73-40-5
  • Formel:C₅H₅N₅O
  • MDL nummer:MFCD00071533
  • Molecular Weight:151.13 g/mol
  • Merck Index:14,4564
  • Smeltende Pt:>300 °C
  • Tæthed:1,67 g/cm³ (20 °C)
  • Opbevaringstemperatur:Køleskab
  • EINECS:200-799-8
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer