Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
1,3-Propansulton ≥99%

1,3-Propansulton ≥99%

Glass bottle for solids, 50 g
Catalog # A11923.18
Toxic
Health
1,3-Propansulton ≥99%
Catalog # A11923.18
CAS Number:  1120-71-4
1,3-Propansulton ≥99%
1,3-Propansulton ≥99%

Glass bottle for solids, 50 g

CAS Number:  1120-71-4
Price
1.810,00 kr
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 99% CAS No.: 1120-71-4 Molecular Formula: C3H6O3S Formula Weight: 122.14 Flash Point: 110°(230°F) Melting Point: 30-33° Boiling Point: 179-181°/30mm Beilstein Reference: 109782 UN No.: 2811 MDL No.: MFCD00005355

Synonyms:  1,2-Oxathiolane 2,2-dioxide
  • CAS Number:1120-71-4
  • Formula:C₃H₆O₃S
  • MDL Number:MFCD00005355
  • Molecular Weight:122.14 g/mol
  • Boiling Pt:179…181 °C (30 mmHg)
  • Melting Pt:30…33 °C
  • Density:1,392
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Stue temperatur
  • EINECS:214-317-9
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 g

Specifications