Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

1,3-Propansulton ≥99%

Glass bottle for solids, 10 g
Katalog # A11923.09
Giftig
Sundhed
Leverandør:   Thermo Fisher Scientific
1,3-Propansulton ≥99%
Katalog # A11923.09
CAS nummer:  1120-71-4
1,3-Propansulton ≥99%
1,3-Propansulton ≥99%

Glass bottle for solids, 10 g

CAS nummer:  1120-71-4
Price
kr580.00
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents

Purity: 99% CAS No.: 1120-71-4 Molecular Formula: C3H6O3S Formula Weight: 122.14 Flash Point: 110°(230°F) Melting Point: 30-33° Boiling Point: 179-181°/30mm Beilstein Reference: 109782 UN No.: 2811 MDL No.: MFCD00005355

Synonymer:  1,2-Oxathiolane 2,2-dioxide
  • CAS nummer:1120-71-4
  • Formel:C₃H₆O₃S
  • MDL nummer:MFCD00005355
  • Molecular Weight:122.14 g/mol
  • Kogende Pt:179…181 °C (30 mmHg)
  • Smeltende Pt:30…33 °C
  • Tæthed:1,392
  • Flash Pt:>110 °C (230 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:214-317-9
  • FN:2811
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifikationer