Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
1-Naphtol ≥99%

1-Naphtol ≥99%

Glass bottle for solids, 500 g
Katalog # A11862.36
Ætsende
Irriterende
Leverandør:   Thermo Fisher Scientific
1-Naphtol ≥99%
Katalog # A11862.36
CAS nummer:  90-15-3
1-Naphtol ≥99%
1-Naphtol ≥99%

Glass bottle for solids, 500 g

CAS nummer:  90-15-3
Price
824,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 90-15-3 Molecular Formula: C10H8O Formula Weight: 144.17 Flash Point: 125°(257°F) Odour: schwach, phenolartig Boiling Point: 278-280° Solubility: Soluble in alcohol, benzene, chloroform, ether Beilstein Reference: 1817321 Merck Reference: 14,6383 UN No.: 2811 MDL No.: MFCD00003930

Synonymer:  α-Naphtol
  • CAS nummer:90-15-3
  • Formel:C₁₀H₈O
  • MDL nummer:MFCD00003930
  • Molecular Weight:144.17 g/mol
  • Merck Index:14,6383
  • Kogende Pt:278…280 °C
  • Smeltende Pt:95…97 °C
  • Tæthed:1,28
  • Flash Pt:125 °C (257 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-969-4
  • FN:2811
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g
  • Cat. No.:
    A11862.36

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.