trans-4-Hydroxy-L(-)-prolin ≥99%
Glass bottle for solids, 100 g
Leverandør: Thermo Fisher Scientific
trans-4-Hydroxy-L(-)-prolin ≥99%
Katalog # A11851.22
Leverandør: Thermo Fisher Scientific
CAS nummer: 51-35-4
Gå til:
Product Details & Documents
Purity: 99+% CAS No.: 51-35-4 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: 270-273° dec. Beilstein Reference: 471933 Merck Reference: 14,4840 Optical Rotation: -76° (c=4 in water) MDL No.: MFCD00064320
Synonymer: L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
- CAS nummer:51-35-4
- Formel:C₅H₉NO₃
- MDL nummer:MFCD00064320
- Molecular Weight:131.13 g/mol
- Merck Index:14,4840
- Kogende Pt:296 °C (1013 hPa)
- Smeltende Pt:270…273 °C (dec.)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-091-9
Documents
Certifikater
Søg efter partinummerALFAA11851.09_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



