trans-4-Hydroxy-L(-)-prolin ≥99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
trans-4-Hydroxy-L(-)-prolin ≥99%
Catalog # A11851.14
Supplier: Thermo Fisher Scientific
CAS Number: 51-35-4
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 51-35-4 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: 270-273° dec. Beilstein Reference: 471933 Merck Reference: 14,4840 Optical Rotation: -76° (c=4 in water) MDL No.: MFCD00064320
Synonyms: L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
- CAS Number:51-35-4
- Formula:C₅H₉NO₃
- MDL Number:MFCD00064320
- Molecular Weight:131.13 g/mol
- Merck Index:14,4840
- Boiling Pt:296 °C (1013 hPa)
- Melting Pt:270…273 °C (dec.)
- Storage Temperature:Stue temperatur
- EINECS:200-091-9
Documents
Certificates
Search by Lot NumberALFAA11851.09_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



