Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
trans-4-Hydroxy-L(-)-prolin ≥99%

trans-4-Hydroxy-L(-)-prolin ≥99%

Glass bottle for solids, 10 g
Katalog # A11851.09
Leverandør:   Thermo Fisher Scientific
trans-4-Hydroxy-L(-)-prolin ≥99%
Katalog # A11851.09
CAS nummer:  51-35-4
trans-4-Hydroxy-L(-)-prolin ≥99%
trans-4-Hydroxy-L(-)-prolin ≥99%

Glass bottle for solids, 10 g

CAS nummer:  51-35-4
Price
608,00 kr
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents

Purity: 99+% CAS No.: 51-35-4 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: 270-273° dec. Beilstein Reference: 471933 Merck Reference: 14,4840 Optical Rotation: -76° (c=4 in water) MDL No.: MFCD00064320

Synonymer:  L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
  • CAS nummer:51-35-4
  • Formel:C₅H₉NO₃
  • MDL nummer:MFCD00064320
  • Molecular Weight:131.13 g/mol
  • Merck Index:14,4840
  • Kogende Pt:296 °C (1013 hPa)
  • Smeltende Pt:270…273 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-091-9
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.