Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Natriumoctylsulfat ≥99%

Glass bottle for solids, 25 g
Katalog # A11786.14
Irriterende
Leverandør:   Thermo Fisher Scientific
Natriumoctylsulfat ≥99%
Katalog # A11786.14
CAS nummer:  142-31-4
Natriumoctylsulfat ≥99%
Natriumoctylsulfat ≥99%

Glass bottle for solids, 25 g

CAS nummer:  142-31-4
Price
6.160,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 142-31-4 Molecular Formula: CH3(CH2)7OSO3Na Formula Weight: 232.28 Melting Point: ca 195° dec. Beilstein Reference: 3658635

Synonymer:  Octyl sulphate, sodium salt, Octyl sulphuric acid salt
  • CAS nummer:142-31-4
  • Formel:C₈H₁₇NaO₄S
  • MDL nummer:MFCD00007470
  • Molecular Weight:232.28 g/mol
  • Smeltende Pt:∼195 °C (dec.)
  • Tæthed:1,266 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:205-535-5
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer