Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
1,2,4-Triazol ≥99%

1,2,4-Triazol ≥99%

Glass bottle for solids, 100 g
Katalog # A11597.22
Sundhed
Irriterende
Leverandør:   Thermo Fisher Scientific
1,2,4-Triazol ≥99%
Katalog # A11597.22
CAS nummer:  288-88-0
1,2,4-Triazol ≥99%
1,2,4-Triazol ≥99%

Glass bottle for solids, 100 g

CAS nummer:  288-88-0
Price
1.410,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 288-88-0 Molecular Formula: C2H3N3 Formula Weight: 69.07 Flash Point: 158°(316°F) Melting Point: 118-122° Boiling Point: 260° Beilstein Reference: 104767 Merck Reference: 14,9605 MDL No.: MFCD00005228

Synonymer:  1,2,4-1H-Triazole, 1H-1,2,4-Triazole
  • CAS nummer:288-88-0
  • Formel:C₂H₃N₃
  • MDL nummer:MFCD00005228
  • Molecular Weight:69.07 g/mol
  • Merck Index:14,9605
  • Kogende Pt:260 °C
  • Smeltende Pt:118…122 °C
  • Tæthed:1,394 g/cm³ (25 °C)
  • Flash Pt:158 °C (316 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:206-022-9
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.