Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
1,2,4-Triazol ≥99%

1,2,4-Triazol ≥99%

Glass bottle for solids, 25 g
Katalog # A11597.14
Sundhed
Irriterende
Leverandør:   Thermo Fisher Scientific
1,2,4-Triazol ≥99%
Katalog # A11597.14
CAS nummer:  288-88-0
1,2,4-Triazol ≥99%
1,2,4-Triazol ≥99%

Glass bottle for solids, 25 g

CAS nummer:  288-88-0
Price
494,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 288-88-0 Molecular Formula: C2H3N3 Formula Weight: 69.07 Flash Point: 158°(316°F) Melting Point: 118-122° Boiling Point: 260° Beilstein Reference: 104767 Merck Reference: 14,9605 MDL No.: MFCD00005228

Synonymer:  1,2,4-1H-Triazole, 1H-1,2,4-Triazole
  • CAS nummer:288-88-0
  • Formel:C₂H₃N₃
  • MDL nummer:MFCD00005228
  • Molecular Weight:69.07 g/mol
  • Merck Index:14,9605
  • Kogende Pt:260 °C
  • Smeltende Pt:118…122 °C
  • Tæthed:1,394 g/cm³ (25 °C)
  • Flash Pt:158 °C (316 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:206-022-9
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer