Ethylenglycol ≥99%
Glass bottle for solids, 250 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 99% CAS No.: 107-21-1 Molecular Formula: C2H6O2 Formula Weight: 62.07 Flash Point: 119°(246°F) Refractive Index: 1.4310 Odour: Sweet Melting Point: -13° Boiling Point: 196-198° Beilstein Reference: 505945 Merck Reference: 14,3798 MDL No.: MFCD00002885
Synonymer: 1,2-Ethandiol, Glykol
- CAS nummer:107-21-1
- Formel:HOCH₂CH₂OH
- MDL nummer:MFCD00002885
- Molecular Weight:62.07 g/mol
- Merck Index:14,3798
- Kogende Pt:196…198 °C
- Smeltende Pt:–13 °C
- Tæthed:1,113
- Flash Pt:119 °C (246 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-473-3
- NÅ:01-2119456816-28
Documents
Certifikater
Søg efter partinummerALFAA11591.30_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:250 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



