Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

ACES ≥99%

Glass bottle for solids, 100 g
Katalog # A11553.22
Leverandør:   Thermo Fisher Scientific
ACES ≥99%
Katalog # A11553.22
CAS nummer:  7365-82-4
ACES ≥99%
ACES ≥99%

Glass bottle for solids, 100 g

CAS nummer:  7365-82-4
Price
2.100,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 7365-82-4 Molecular Formula: H2NCOCH2NHCH2CH2SO3H Formula Weight: 182.20 Melting Point: ca 275° dec. Beilstein Reference: 2253770 Merck Reference: 14,36 MDL No.: MFCD00008030

Synonymer:  ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid
  • CAS nummer:7365-82-4
  • Formel:H₂NCOCH₂NHCH₂CH₂SO₃H
  • MDL nummer:MFCD00008030
  • Molecular Weight:182.2 g/mol
  • Merck Index:14,36
  • Smeltende Pt:∼ 275 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:230-908-4
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.