Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ethyl sorbate ≥97%

Ethyl sorbate ≥97%

Glass bottle for solids, 100 g
Katalog # A11431.22
Leverandør:   Thermo Fisher Scientific
Ethyl sorbate ≥97%
Katalog # A11431.22
CAS nummer:  2396-84-1
Ethyl sorbate ≥97%
Ethyl sorbate ≥97%

Glass bottle for solids, 100 g

CAS nummer:  2396-84-1
Price
966,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 2396-84-1 Molecular Formula: CH3CH=CHCH=CHCO2CH2CH3 Formula Weight: 140.18 Flash Point: 69°(156°F) Refractive Index: 1.4950 Boiling Point: 195-196° Beilstein Reference: 1362941 MDL No.: MFCD00009296

Synonymer:  Ethyl trans,trans-hexa-2,4-dienoate, (2E,4E)-Ethyl hexa-2,4-dienoate, Ethyl (2E,4E)-hexa-2,4-dienoate, 2,4-Hexadienoic acid ethyl ester, Sorbic acid ethyl ester, (2E,4E)-2,4-Hexadienoic acid ethyl ester, trans,trans-2,4-Hexadienoic acid ethyl ester, (E,E)-2,4-Hexadienoic acid ethyl ester, Ethyl (E,E)-hexa-2,4-dienoate
  • CAS nummer:2396-84-1
  • Formel:C₈H₁₂O₂
  • MDL nummer:MFCD00009296
  • Molecular Weight:140.18 g/mol
  • Kogende Pt:195…196 °C
  • Tæthed:0,948
  • Flash Pt:69 °C (156 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:219-258-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.