Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ethyl sorbate ≥97%

Ethyl sorbate ≥97%

Glass bottle for solids, 25 g
Katalog # A11431.14
Leverandør:   Thermo Fisher Scientific
Ethyl sorbate ≥97%
Katalog # A11431.14
CAS nummer:  2396-84-1
Ethyl sorbate ≥97%
Ethyl sorbate ≥97%

Glass bottle for solids, 25 g

CAS nummer:  2396-84-1
Price
316,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 2396-84-1 Molecular Formula: CH3CH=CHCH=CHCO2CH2CH3 Formula Weight: 140.18 Flash Point: 69°(156°F) Refractive Index: 1.4950 Boiling Point: 195-196° Beilstein Reference: 1362941 MDL No.: MFCD00009296

Synonymer:  Ethyl trans,trans-hexa-2,4-dienoate, (2E,4E)-Ethyl hexa-2,4-dienoate, Ethyl (2E,4E)-hexa-2,4-dienoate, 2,4-Hexadienoic acid ethyl ester, Sorbic acid ethyl ester, (2E,4E)-2,4-Hexadienoic acid ethyl ester, trans,trans-2,4-Hexadienoic acid ethyl ester, (E,E)-2,4-Hexadienoic acid ethyl ester, Ethyl (E,E)-hexa-2,4-dienoate
  • CAS nummer:2396-84-1
  • Formel:C₈H₁₂O₂
  • MDL nummer:MFCD00009296
  • Molecular Weight:140.18 g/mol
  • Kogende Pt:195…196 °C
  • Tæthed:0,948
  • Flash Pt:69 °C (156 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:219-258-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer