Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cyclohexen ≥99%

Cyclohexen ≥99%

Glass bottle, 500 mL
Katalog # A11359.AP
Brandfarlig
Sundhed
Irriterende
Miljø
Leverandør:   Thermo Fisher Scientific
Cyclohexen ≥99%
Katalog # A11359.AP
CAS nummer:  110-83-8
Cyclohexen ≥99%
Cyclohexen ≥99%

Glass bottle, 500 mL

CAS nummer:  110-83-8
Price
471,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

Purity: 99% CAS No.: 110-83-8 Molecular Formula: C6H10 Formula Weight: 82.15 Flash Point: -20°(-4°F) Refractive Index: 1.4460 Odour: Petrol-like Melting Point: -104° Boiling Point: 83-85° Beilstein Reference: 906737 Merck Reference: 14,2727 UN No.: 2256 MDL No.: MFCD00001539

  • CAS nummer:110-83-8
  • Formel:C₆H₁₀
  • MDL nummer:MFCD00001539
  • Molecular Weight:82.15 g/mol
  • Merck Index:14,2727
  • Kogende Pt:83…85 °C
  • Smeltende Pt:–104 °C
  • Tæthed:0,811
  • Flash Pt:–20 °C (–4 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-807-8
  • FN:2256
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL

Specifikationer