1-Octen ≥97%
Glass bottle, 500 mL
Purity: 97+% CAS No.: 111-66-0 Molecular Formula: C8H16 Formula Weight: 112.22 Flash Point: 21°(70°F) Refractive Index: 1.4090 Odour: Petroleum-like Melting Point: -102° Boiling Point: 122-123° Beilstein Reference: 1734497 Merck Reference: 14,1764 UN No.: 3295 MDL No.: MFCD00009548
- :111-66-0
- :C₈H₁₆
- :MFCD00009548
- :112.22 g/mol
- :14,1764
- :122…123 °C
- :–102 °C
- :0,715
- :21 °C (70 °F)
- :Stue temperatur
- :203-893-7
- :3295
- :3,II
Documents
- Pakning:Glass bottle
- Pakn:500 mL