Orders LinkContactUsLinkComponent
1-Octen ≥97%
1-Octen ≥97%
Glass bottle, 500 mL
  A11146.AP
1-Octen ≥97%
  A11146.AP
 :  111-66-0
1-Octen ≥97%
1-Octen ≥97%
Glass bottle, 500 mL
 :  111-66-0
462,00 kr
1 * 500 mL
Purity: 97+% CAS No.: 111-66-0 Molecular Formula: C8H16 Formula Weight: 112.22 Flash Point: 21°(70°F) Refractive Index: 1.4090 Odour: Petroleum-like Melting Point: -102° Boiling Point: 122-123° Beilstein Reference: 1734497 Merck Reference: 14,1764 UN No.: 3295 MDL No.: MFCD00009548
  •  :111-66-0
  •  :C₈H₁₆
  •  :MFCD00009548
  •  :112.22 g/mol
  •  :14,1764
  •  :122…123 °C
  •  :–102 °C
  •  :0,715
  •  :21 °C (70 °F)
  •  :Stue temperatur
  •  :203-893-7
  •  :3295
  •  :3,II
  • Pakning:
    Glass bottle
  • Pakn:
    500 mL