Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

1-Octen ≥97%

Glass bottle, 2.500 mL
Katalog # A11146.0F
Brandfarlig
Sundhed
Irriterende
Leverandør:   Thermo Fisher Scientific
1-Octen ≥97%
Katalog # A11146.0F
CAS nummer:  111-66-0
1-Octen ≥97%
1-Octen ≥97%

Glass bottle, 2.500 mL

CAS nummer:  111-66-0
Price
1.490,00 kr
Log ind for tilgængelighed
1 * 2.500 mL
Product Details & Documents

Purity: 97+% CAS No.: 111-66-0 Molecular Formula: C8H16 Formula Weight: 112.22 Flash Point: 21°(70°F) Refractive Index: 1.4090 Odour: Petroleum-like Melting Point: -102° Boiling Point: 122-123° Beilstein Reference: 1734497 Merck Reference: 14,1764 UN No.: 3295 MDL No.: MFCD00009548

  • CAS nummer:111-66-0
  • Formel:C₈H₁₆
  • MDL nummer:MFCD00009548
  • Molecular Weight:112.22 g/mol
  • Merck Index:14,1764
  • Kogende Pt:122…123 °C
  • Smeltende Pt:–102 °C
  • Tæthed:0,715
  • Flash Pt:21 °C (70 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-893-7
  • FN:3295
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2.500 mL

Specifikationer