(4-Nitrophenyl)eddikesyreethylester ≥98%
Glass bottle for solids, 25 g
Purity: 98% CAS No.: 5445-26-1 Molecular Formula: C10H11NO4 Formula Weight: 209.20 Melting Point: 62-66° Beilstein Reference: 2053241
: Ethyl 4-nitrohenylacetate, Ethyl (4-nitrophenyl)acetate, Ethyl 4-nitrophenylacetate
- :5445-26-1
- :C₁₀H₁₁NO₄
- :MFCD00017046
- :209.2 g/mol
- :62…66 °C
- :Stue temperatur
- :226-646-5
- Pakning:Glass bottle for solids
- Pakn:25 g