Orders LinkContactUsLinkComponent
(4-Nitrophenyl)eddikesyreethylester ≥98%
Glass bottle for solids, 25 g
  A11101.14
(4-Nitrophenyl)eddikesyreethylester ≥98%
  A11101.14
 :  5445-26-1
(4-Nitrophenyl)eddikesyreethylester ≥98%
(4-Nitrophenyl)eddikesyreethylester ≥98%
Glass bottle for solids, 25 g
 :  5445-26-1
2.020,00 kr
1 * 25 g
Purity: 98% CAS No.: 5445-26-1 Molecular Formula: C10H11NO4 Formula Weight: 209.20 Melting Point: 62-66° Beilstein Reference: 2053241
 :  Ethyl 4-nitrohenylacetate, Ethyl (4-nitrophenyl)acetate, Ethyl 4-nitrophenylacetate
  •  :5445-26-1
  •  :C₁₀H₁₁NO₄
  •  :MFCD00017046
  •  :209.2 g/mol
  •  :62…66 °C
  •  :Stue temperatur
  •  :226-646-5
  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g