Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Barbitursyre ≥99%

Barbitursyre ≥99%

Glass bottle for solids, 500 g
Katalog # A10934.36
Leverandør:   Thermo Fisher Scientific
Barbitursyre ≥99%
Katalog # A10934.36
CAS nummer:  67-52-7
Barbitursyre ≥99%
Barbitursyre ≥99%

Glass bottle for solids, 500 g

CAS nummer:  67-52-7
Price
1.410,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 67-52-7 Molecular Formula: C4H4N2O3 Formula Weight: 128.09 Flash Point: 150°(302°F) pH: 2-3 Melting Point: ca 250° dec. Beilstein Reference: 120502 Merck Reference: 14,963 MDL No.: MFCD00006666

Synonymer:  N,N'-Malonylurea, 2,4,6-Trihydroxypyrimidine
  • CAS nummer:67-52-7
  • Formel:C₄H₄N₂O₃
  • MDL nummer:MFCD00006666
  • Molecular Weight:128.09 g/mol
  • Merck Index:14,963
  • Kogende Pt:441 °C (1013 hPa)
  • Smeltende Pt:∼ 250 °C (dec.)
  • Flash Pt:150 °C (302 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-658-0
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.