Barbitursyre ≥99%
Glass bottle for solids, 100 g
Gå til:
Discontinued
Produktet er udgået. Men vi fandt lignende alternative produkter.
Alternativer
BARBITURIC ACID 99% 1 * 500 g Katalog #A10934.36
Produktet A10934.36 er en erstatning for A10934.22, men varierer efter mærke, emballage eller mængde.
Product Details & Documents
Purity: 99% CAS No.: 67-52-7 Molecular Formula: C4H4N2O3 Formula Weight: 128.09 Flash Point: 150°(302°F) pH: 2-3 Melting Point: ca 250° dec. Beilstein Reference: 120502 Merck Reference: 14,963 MDL No.: MFCD00006666
Synonymer: N,N'-Malonylurea, 2,4,6-Trihydroxypyrimidine
- CAS nummer:67-52-7
- Formel:C₄H₄N₂O₃
- MDL nummer:MFCD00006666
- Molecular Weight:128.09 g/mol
- Merck Index:14,963
- Kogende Pt:441 °C (1013 hPa)
- Smeltende Pt:∼ 250 °C (dec.)
- Flash Pt:150 °C (302 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-658-0
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



