Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Benzophenon ≥99%

Glass bottle for solids, 5.000 g
Katalog # A10739.0I
Miljø
Leverandør:   Thermo Fisher Scientific
Benzophenon ≥99%
Katalog # A10739.0I
CAS nummer:  119-61-9
Benzophenon ≥99%
Benzophenon ≥99%

Glass bottle for solids, 5.000 g

CAS nummer:  119-61-9
Price
3.260,00 kr
Log ind for tilgængelighed
1 * 5.000 g
Product Details & Documents

Purity: 99% CAS No.: 119-61-9 Molecular Formula: C13H10O Formula Weight: 182.22 Flash Point: 138°(280°F) Physical Form: Cryst flakes or powder Density: 1.111 Appearance: White Odour: Pleasant Melting Point: 47-50° Boiling Point: 305° Beilstein Reference: 1238185 Merck Reference: 14,1098 UN No.: 3077 MDL No.: MFCD00003076

Synonymer:  Diphenylmethanone, Diphenyl ketone
  • CAS nummer:119-61-9
  • Formel:C₁₃H₁₀O
  • MDL nummer:MFCD00003076
  • Molecular Weight:182.22 g/mol
  • Merck Index:14,1098
  • Kogende Pt:305 °C
  • Smeltende Pt:47…50 °C
  • Tæthed:1,111
  • Flash Pt:138 °C (280 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-337-6
  • FN:3077
  • ADR:9,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5.000 g

Specifikationer