Orders LinkContactUsLinkComponent
EDTA 99%
Glass bottle for solids, 500 g
Katalog # A10713.36
Irriterende
Leverandør:   Thermo Fisher Scientific
EDTA 99%
Katalog # A10713.36
CAS nummer:  60-00-4
EDTA 99%
EDTA 99%
Glass bottle for solids, 500 g
CAS nummer:  60-00-4
Price
kr741.00
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Purity: 99% CAS No.: 60-00-4 Molecular Formula: C10H16N2O8 Formula Weight: 292.24 Melting Point: ca 245° dec. Beilstein Reference: 1716295 Merck Reference: 14,3517 MDL No.: MFCD00003541
Synonymer:  Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
  • CAS nummer:60-00-4
  • Formel:C₁₀H₁₆N₂O₈
  • MDL nummer:MFCD00003541
  • Molecular Weight:292.25 g/mol
  • Merck Index:14,3517
  • Kogende Pt:614 °C (1013 hPa)
  • Smeltende Pt:∼ 245 °C (dec.)
  • Tæthed:0,86 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-449-4
  • NÅ:01-2119486399-18
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g