EDTA 99%
Glass bottle for solids, 250 g
Purity: 99% CAS No.: 60-00-4 Molecular Formula: C10H16N2O8 Formula Weight: 292.24 Melting Point: ca 245° dec. Beilstein Reference: 1716295 Merck Reference: 14,3517 MDL No.: MFCD00003541
: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
- :60-00-4
- :C₁₀H₁₆N₂O₈
- :MFCD00003541
- :292.25 g/mol
- :14,3517
- :614 °C (1013 hPa)
- :∼ 245 °C (dec.)
- :0,86 g/cm³ (20 °C)
- :>100 °C
- :Stue temperatur
- :200-449-4
- :01-2119486399-18
- Pakning:Glass bottle for solids
- Pakn:250 g