Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
EDTA 99%

EDTA 99%

Glass bottle for solids, 5.000 g
Katalog # A10713.0I
Irriterende
Leverandør:   Thermo Fisher Scientific
EDTA 99%
Katalog # A10713.0I
CAS nummer:  60-00-4
EDTA 99%
EDTA 99%

Glass bottle for solids, 5.000 g

CAS nummer:  60-00-4
Price
3.400,00 kr
Log ind for tilgængelighed
1 * 5.000 g
Product Details & Documents

Purity: 99% CAS No.: 60-00-4 Molecular Formula: C10H16N2O8 Formula Weight: 292.24 Melting Point: ca 245° dec. Beilstein Reference: 1716295 Merck Reference: 14,3517 MDL No.: MFCD00003541

Synonymer:  Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
  • CAS nummer:60-00-4
  • Formel:C₁₀H₁₆N₂O₈
  • MDL nummer:MFCD00003541
  • Molecular Weight:292.25 g/mol
  • Merck Index:14,3517
  • Kogende Pt:614 °C (1013 hPa)
  • Smeltende Pt:∼ 245 °C (dec.)
  • Tæthed:0,86 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-449-4
  • NÅ:01-2119486399-18
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5.000 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.