D(+)-Xylose ≥98%
Glass bottle for solids, 500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 98+% CAS No.: 58-86-6 Molecular Formula: C5H10O5 Formula Weight: 150.13 Flash Point: 100°(212°F) Physical Form: Cryst powder Density: 1.525 Sensitivity: Hygroscopic Appearance: White Melting Point: 147-151° Beilstein Reference: 1562108 Merck Reference: 14,10087 Optical Rotation: +19° (c=10 in water + NH4OH) MDL No.: MFCD00151475
- CAS nummer:58-86-6
- Formel:C₅H₁₀O₅
- MDL nummer:MFCD00151475
- Molecular Weight:150.13 g/mol
- Merck Index:14,10087
- Kogende Pt:331 °C (1013 hPa)
- Smeltende Pt:147…151 °C
- Tæthed:1,525
- Flash Pt:>100 °C (212 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-400-7
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g



