Nitrobenzen ≥99%
Glass bottle for solids, 500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 99% CAS No.: 98-95-3 Molecular Formula: C6H5NO2 Formula Weight: 123.11 Flash Point: 87°(188°F) Refractive Index: 1.5520 Melting Point: 5-6° Boiling Point: 210-212° Beilstein Reference: 507540 Merck Reference: 14,6588 UN No.: 1662 MDL No.: MFCD00007043
- CAS nummer:98-95-3
- Formel:C₆H₅NO₂
- MDL nummer:MFCD00007043
- Molecular Weight:123.11 g/mol
- Merck Index:14,6588
- Kogende Pt:210…212 °C
- Smeltende Pt:5…6 °C
- Tæthed:1,196
- Flash Pt:87 °C (188 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-716-0
- FN:1662
- ADR:6.1,II
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g



