Orders LinkContactUsLinkComponent
Nitrobenzen ≥99%
Glass bottle for solids, 2.500 g
  A10585.0E
Nitrobenzen ≥99%
  A10585.0E
 :  98-95-3
Nitrobenzen ≥99%
Nitrobenzen ≥99%
Glass bottle for solids, 2.500 g
 :  98-95-3
870,00 kr
1 * 2.500 g
Purity: 99% CAS No.: 98-95-3 Molecular Formula: C6H5NO2 Formula Weight: 123.11 Flash Point: 87°(188°F) Refractive Index: 1.5520 Melting Point: 5-6° Boiling Point: 210-212° Beilstein Reference: 507540 Merck Reference: 14,6588 UN No.: 1662 MDL No.: MFCD00007043
  •  :98-95-3
  •  :C₆H₅NO₂
  •  :MFCD00007043
  •  :123.11 g/mol
  •  :14,6588
  •  :210…212 °C
  •  :5…6 °C
  •  :1,196
  •  :87 °C (188 °F)
  •  :Stue temperatur
  •  :202-716-0
  •  :1662
  •  :6.1,II
  • Pakning:
    Glass bottle for solids
  • Pakn:
    2.500 g