Iodmethan ≥99% stabilised
Glass bottle for solids, 50 g
Stabilized with copper
Purity: 99% CAS No.: 74-88-4 Molecular Formula: CH3I Formula Weight: 141.94 Storage Temperature: 2°C to 8°C Refractive Index: 1.5300 Odour: Pungent Melting Point: -67° Boiling Point: 41-43° Beilstein Reference: 969135 Merck Reference: 14,6087 UN No.: 2644 MDL No.: MFCD00001073
: Methyliodid
- :74-88-4
- :CH₃I
- :MFCD00001073
- :141.94 g/mol
- :14,6087
- :41…43 °C
- :–67 °C
- :2,28
- :Køleskab
- :200-819-5
- :6.1,I
- :Stabilised with copper
- :2644
- Pakning:Glass bottle for solids
- Pakn:50 g



