Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Iodmethan ≥99% stabilised

Iodmethan ≥99% stabilised

Glass bottle for solids, 1.000 g
Catalog # A10559.0B
Toxic
Health
Irritant
Iodmethan ≥99% stabilised
Catalog # A10559.0B
CAS Number:  74-88-4
Iodmethan ≥99% stabilised
Iodmethan ≥99% stabilised

Glass bottle for solids, 1.000 g

CAS Number:  74-88-4
Price
5.710,00 kr
Log in for availability
1 * 1.000 g
Product Details & Documents

Stabilized with copper

Purity: 99% CAS No.: 74-88-4 Molecular Formula: CH3I Formula Weight: 141.94 Storage Temperature: 2°C to 8°C Refractive Index: 1.5300 Odour: Pungent Melting Point: -67° Boiling Point: 41-43° Beilstein Reference: 969135 Merck Reference: 14,6087 UN No.: 2644 MDL No.: MFCD00001073

Synonyms:  Methyliodid
  • CAS Number:74-88-4
  • Formula:CH₃I
  • MDL Number:MFCD00001073
  • Molecular Weight:141.94 g/mol
  • Merck Index:14,6087
  • Boiling Pt:41…43 °C
  • Melting Pt:–67 °C
  • Density:2,28
  • Storage Temperature:Køleskab
  • EINECS:200-819-5
  • ADR:6.1,I
  •  :Stabilised with copper
  • UN:2644
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1.000 g

Specifications