4'-Hydroxy-3'-methoxyacetophenon ≥98%
Glass bottle for solids, 250 g
Leverandør: Thermo Fisher Scientific
4'-Hydroxy-3'-methoxyacetophenon ≥98%
Katalog # A10439.30
Leverandør: Thermo Fisher Scientific
CAS nummer: 498-02-2
Gå til:
Product Details & Documents
Purity: 98% CAS No.: 498-02-2 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 112-116° Boiling Point: 295-300° Merck Reference: 14,741 MDL No.: MFCD00008747
Synonymer: 4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin
- CAS nummer:498-02-2
- Formel:C₉H₁₀O₃
- MDL nummer:MFCD00008747
- Molecular Weight:166.18 g/mol
- Merck Index:14,741
- Kogende Pt:295…300 °C
- Smeltende Pt:112…116 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:207-854-5
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:250 g



