Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Citronsyre ≥99%

Citronsyre ≥99%

Glass bottle for solids, 250 g
Katalog # A10395.30
Ætsende
Irriterende
Leverandør:   Thermo Fisher Scientific
Citronsyre ≥99%
Katalog # A10395.30
CAS nummer:  77-92-9
Citronsyre ≥99%
Citronsyre ≥99%

Glass bottle for solids, 250 g

CAS nummer:  77-92-9
Price
359,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents

Purity: 99+% CAS No.: 77-92-9 Molecular Formula: C6H8O7 Formula Weight: 192.13 Odour: Odourless pH: 1,6 Melting Point: 153-154° Beilstein Reference: 782061 Merck Reference: 14,2326 MDL No.: MFCD00011669

Synonymer:  2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
  • CAS nummer:77-92-9
  • Formel:HOC(COOH)(CH₂COOH)₂
  • MDL nummer:MFCD00011669
  • Molecular Weight:192.13 g/mol
  • Merck Index:14,2326
  • Kogende Pt:310 °C (1013 hPa)
  • Smeltende Pt:153…154 °C
  • Tæthed:1,665
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-069-1
  • NÅ:01-2119457026-42-0000
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.