Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

4-Ethylanisol ≥98%

Glass bottle for solids, 50 g
Katalog # A10391.18
Leverandør:   Thermo Fisher Scientific
4-Ethylanisol ≥98%
Katalog # A10391.18
CAS nummer:  1515-95-3
4-Ethylanisol ≥98%
4-Ethylanisol ≥98%

Glass bottle for solids, 50 g

CAS nummer:  1515-95-3
Price
2.150,00 kr
Log ind for tilgængelighed
1 * 50 g
Product Details & Documents

Purity: 98+% CAS No.: 1515-95-3 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 79°(174°F) Refractive Index: 1.5085 Boiling Point: 195° Beilstein Reference: 774365 MDL No.: MFCD00015176

Synonymer:  1-Ethyl-4-methoxybenzene
  • CAS nummer:1515-95-3
  • Formel:C₉H₁₂O
  • MDL nummer:MFCD00015176
  • Molecular Weight:136.19 g/mol
  • Kogende Pt:195 °C
  • Tæthed:0,962
  • Flash Pt:79 °C (174 °F)
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 g

Specifikationer