Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
4-Ethylanisol ≥98%

4-Ethylanisol ≥98%

Glass bottle for solids, 10 g
Catalog # A10391.09
4-Ethylanisol ≥98%
Catalog # A10391.09
CAS Number:  1515-95-3
4-Ethylanisol ≥98%
4-Ethylanisol ≥98%

Glass bottle for solids, 10 g

CAS Number:  1515-95-3
Price
599,00 kr
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 98+% CAS No.: 1515-95-3 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 79°(174°F) Refractive Index: 1.5085 Boiling Point: 195° Beilstein Reference: 774365 MDL No.: MFCD00015176

Synonyms:  1-Ethyl-4-methoxybenzene
  • CAS Number:1515-95-3
  • Formula:C₉H₁₂O
  • MDL Number:MFCD00015176
  • Molecular Weight:136.19 g/mol
  • Boiling Pt:195 °C
  • Density:0,962
  • Flash Pt:79 °C (174 °F)
  • Storage Temperature:Stue temperatur
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifications