Orders LinkContactUsLinkComponent
Morpholin ≥99%
Morpholin ≥99%
Glass bottle for solids, 2.500 g
Katalog # A10355.0E
Ætsende
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
Morpholin ≥99%
Katalog # A10355.0E
CAS nummer:  110-91-8
Morpholin ≥99%
Morpholin ≥99%
Glass bottle for solids, 2.500 g
CAS nummer:  110-91-8
Price
1.160,00 kr
Log ind for tilgængelighed
1 * 2.500 g
Product Details & Documents
Purity: 99% CAS No.: 110-91-8 Molecular Formula: C4H9NO Formula Weight: 87.12 Flash Point: 35°(95°F) Physical Form: Liquid Density: 1.007 Refractive Index: 1.4540 Sensitivity: Hygroscopic Appearance: Colourless Odour: Amine-like Melting Point: -5° Boiling Point: 129° Beilstein Reference: 102549 Merck Reference: 14,6277 UN No.: 2054 MDL No.: MFCD00005972
  • CAS nummer:110-91-8
  • Formel:C₄H₉NO
  • MDL nummer:MFCD00005972
  • Molecular Weight:87.12 g/mol
  • Merck Index:14,6277
  • Kogende Pt:129 °C
  • Smeltende Pt:–5 °C
  • Tæthed:1,007
  • Flash Pt:35 °C (95 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-815-1
  • FN:2054
  • ADR:8,I
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    2.500 g
Specifikationer