Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
L-Methionin ≥98%

L-Methionin ≥98%

Glass bottle for solids, 500 g
Katalog # A10318.36
Leverandør:   Thermo Fisher Scientific
L-Methionin ≥98%
Katalog # A10318.36
CAS nummer:  63-68-3
L-Methionin ≥98%
L-Methionin ≥98%

Glass bottle for solids, 500 g

CAS nummer:  63-68-3
Price
3.160,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Purity: 98+% CAS No.: 63-68-3 Molecular Formula: C5H11NO2S Formula Weight: 149.21 Melting Point: ca 280° dec. Beilstein Reference: 1722294 Merck Reference: 14,5975 Optical Rotation: +23° (c=2 in 2N HCl) MDL No.: MFCD00063097

Synonymer:  (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
  • CAS nummer:63-68-3
  • Formel:C₅H₁₁NO₂S
  • MDL nummer:MFCD00063097
  • Molecular Weight:149.21 g/mol
  • Merck Index:14,5975
  • Kogende Pt:278 °C (1013 hPa)
  • Smeltende Pt:∼ 280 °C (dec.)
  • Tæthed:1,373 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-562-9
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.