Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
L-Methionin ≥98%

L-Methionin ≥98%

Glass bottle for solids, 100 g
Katalog # A10318.22
Leverandør:   Thermo Fisher Scientific
L-Methionin ≥98%
Katalog # A10318.22
CAS nummer:  63-68-3
L-Methionin ≥98%
L-Methionin ≥98%

Glass bottle for solids, 100 g

CAS nummer:  63-68-3
Price
827,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 98+% CAS No.: 63-68-3 Molecular Formula: C5H11NO2S Formula Weight: 149.21 Melting Point: ca 280° dec. Beilstein Reference: 1722294 Merck Reference: 14,5975 Optical Rotation: +23° (c=2 in 2N HCl) MDL No.: MFCD00063097

Synonymer:  (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
  • CAS nummer:63-68-3
  • Formel:C₅H₁₁NO₂S
  • MDL nummer:MFCD00063097
  • Molecular Weight:149.21 g/mol
  • Merck Index:14,5975
  • Kogende Pt:278 °C (1013 hPa)
  • Smeltende Pt:∼ 280 °C (dec.)
  • Tæthed:1,373 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-562-9
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer