Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

2-Bromethanol ≥97%

Glass bottle for solids, 250 g
Katalog # A10275.30
Giftig
Ætsende
Leverandør:   Thermo Fisher Scientific
2-Bromethanol ≥97%
Katalog # A10275.30
CAS nummer:  540-51-2
2-Bromethanol ≥97%
2-Bromethanol ≥97%

Glass bottle for solids, 250 g

CAS nummer:  540-51-2
Price
1.810,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents

Purity: 97% CAS No.: 540-51-2 Molecular Formula: C2H5BrO Formula Weight: 124.97 Flash Point: 110°(230°F) Refractive Index: 1.4930 Melting Point: -80° Boiling Point: 149-150° Beilstein Reference: 878140 Merck Reference: 14,3792 UN No.: 2922 MDL No.: MFCD00002827

  • CAS nummer:540-51-2
  • Formel:BrH₂CCH₂OH
  • MDL nummer:MFCD00002827
  • Molecular Weight:124.97 g/mol
  • Merck Index:14,3792
  • Kogende Pt:149…150 °C
  • Smeltende Pt:–80 °C
  • Tæthed:1,762
  • Flash Pt:>110 °C (230 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:208-748-1
  • FN:2922
  • ADR:8,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer