Propyl 4-hydroxybenzoat ≥99%
Glass bottle for solids, 5.000 g
Leverandør: Thermo Fisher Scientific
Propyl 4-hydroxybenzoat ≥99%
Katalog # A10180.0I
Leverandør: Thermo Fisher Scientific
CAS nummer: 94-13-3
Gå til:
Product Details & Documents
Purity: 99+% CAS No.: 94-13-3 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 180°(356°F) Melting Point: 95-99° Beilstein Reference: 1103245 Merck Reference: 14,7866 MDL No.: MFCD00002354
Synonymer: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester
- CAS nummer:94-13-3
- Formel:C₁₀H₁₂O₃
- MDL nummer:MFCD00002354
- Molecular Weight:180.2 g/mol
- Merck Index:14,7866
- Kogende Pt:292 °C (1013 hPa)
- Smeltende Pt:95…99 °C
- Tæthed:1,28 g/cm³ (20 °C)
- Flash Pt:180 °C (356 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-307-7
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5.000 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



