Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Propyl 4-hydroxybenzoat ≥99%

Glass bottle for solids, 1.000 g
Katalog # A10180.0B
Irriterende
Leverandør:   Thermo Fisher Scientific
Propyl 4-hydroxybenzoat ≥99%
Katalog # A10180.0B
CAS nummer:  94-13-3
Propyl 4-hydroxybenzoat ≥99%
Propyl 4-hydroxybenzoat ≥99%

Glass bottle for solids, 1.000 g

CAS nummer:  94-13-3
Price
1.550,00 kr
Log ind for tilgængelighed
1 * 1.000 g
Product Details & Documents

Purity: 99+% CAS No.: 94-13-3 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 180°(356°F) Melting Point: 95-99° Beilstein Reference: 1103245 Merck Reference: 14,7866 MDL No.: MFCD00002354

Synonymer:  Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester
  • CAS nummer:94-13-3
  • Formel:C₁₀H₁₂O₃
  • MDL nummer:MFCD00002354
  • Molecular Weight:180.2 g/mol
  • Merck Index:14,7866
  • Kogende Pt:292 °C (1013 hPa)
  • Smeltende Pt:95…99 °C
  • Tæthed:1,28 g/cm³ (20 °C)
  • Flash Pt:180 °C (356 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:202-307-7
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1.000 g

Specifikationer