Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ac-β-Ala-OH 97%

Ac-β-Ala-OH 97%

1 g
Catalog # H50208.03
Ac-β-Ala-OH 97%
Catalog # H50208.03
CAS Number:  3025-95-4
Ac-β-Ala-OH 97%
Ac-β-Ala-OH 97%

1 g

CAS Number:  3025-95-4
Price
520,00 kr
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 97% CAS No.: 3025-95-4 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: 77° MDL No.: MFCD00037296

Synonyms:  N-Ac-β-Ala-OH, 3-Acetamidopropanoic acid, N-Acetyl-beta-alanine, N-Acetyl-b-alanine
  • CAS Number:3025-95-4
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00037296
  • Molecular Weight:131.1 g/mol
  • Melting Pt:77 °C
  • Storage Temperature:Stue temperatur
  • EINECS:221-185-6
Specifications

Specifications

  • Pakn:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.