Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Methylacetat ≥99%

Methylacetat ≥99%

Glass bottle, 2.500 mL
Katalog # L14475.0F
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
Methylacetat ≥99%
Katalog # L14475.0F
CAS nummer:  79-20-9
Methylacetat ≥99%
Methylacetat ≥99%

Glass bottle, 2.500 mL

CAS nummer:  79-20-9
Price
822,00 kr
Log ind for tilgængelighed
1 * 2.500 mL
Product Details & Documents

Purity: 99% CAS No.: 79-20-9 Molecular Formula: CH3COOCH3 Formula Weight: 74.08 Flash Point: -10°(14°F) Refractive Index: 1.3620 Odour: Pleasant Melting Point: -98° Boiling Point: 56-58° Beilstein Reference: 1736662 Merck Reference: 14,6008 UN No.: 1231 MDL No.: MFCD00008711

Synonymer:  Acetic acid methyl ester
  • CAS nummer:79-20-9
  • Formel:C₃H₆O₂
  • MDL nummer:MFCD00008711
  • Molecular Weight:74.08 g/mol
  • Merck Index:14,6008
  • Kogende Pt:56…58 °C
  • Smeltende Pt:–98 °C
  • Tæthed:0,934
  • Flash Pt:–10 °C (14 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-185-2
  • FN:1231
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2.500 mL

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.