Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumcarbonat ≥98%

Natriumcarbonat ≥98%

Glass bottle for solids, 5.000 g
Katalog # L13098.0I
Irriterende
Leverandør:   Thermo Fisher Scientific
Natriumcarbonat ≥98%
Katalog # L13098.0I
CAS nummer:  497-19-8
Natriumcarbonat ≥98%
Natriumcarbonat ≥98%

Glass bottle for solids, 5.000 g

CAS nummer:  497-19-8
Price
1.170,00 kr
Log ind for tilgængelighed
1 * 5.000 g
Product Details & Documents

Purity: 98% CAS No.: 497-19-8 Molecular Formula: Na2CO3 Formula Weight: 105.99 Refractive Index: 1.535 Odour: Odourless pH: 11,5 Beilstein Reference: 4154566 Merck Reference: 14,8596 MDL No.: MFCD00003494

Synonymer:  Calcined soda, Carbonic acid disodium salt
  • CAS nummer:497-19-8
  • Formel:Na₂CO₃
  • MDL nummer:MFCD00003494
  • Molecular Weight:105.99 g/mol
  • Merck Index:14,8596
  • Kogende Pt:1600 °C (1013 hPa)
  • Smeltende Pt:854 °C
  • Tæthed:2,53
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:207-838-8
  • NÅ:01-2119485498-19
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5.000 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.