Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Triethylcitrat ≥99%

Triethylcitrat ≥99%

1.000 g
Katalog # L12639.0B
Leverandør:   Thermo Fisher Scientific
Triethylcitrat ≥99%
Katalog # L12639.0B
CAS nummer:  77-93-0
Triethylcitrat ≥99%
Triethylcitrat ≥99%

1.000 g

CAS nummer:  77-93-0
Price
843,00 kr
Log ind for tilgængelighed
1 * 1.000 g
Product Details & Documents

Purity: 99% CAS No.: 77-93-0 Molecular Formula: C12H20O7 Formula Weight: 276.29 Flash Point: 176°(348°F) Refractive Index: 1.4420 Melting Point: -55° Boiling Point: 126-127°/1mm Beilstein Reference: 1801199 Merck Reference: 14,2326 MDL No.: MFCD00009201

Synonymer:  Triethyl 2-hydroxypropane-1,2,3-tricarboxylate, Hydroxytricarballylic acid triethyl ester, Citric acid triethyl ester
  • CAS nummer:77-93-0
  • Formel:C₁₂H₂₀O₇
  • MDL nummer:MFCD00009201
  • Molecular Weight:276.29 g/mol
  • Merck Index:14,2326
  • Kogende Pt:126…127 °C (1 mmHg)
  • Smeltende Pt:–55 °C
  • Tæthed:1,137
  • Flash Pt:176 °C (348 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-070-7
Specifikationer

Specifikationer

  • Pakn:
    1.000 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.