Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sorbitanmonostearat

Sorbitanmonostearat

Glass bottle for solids, 1.000 g
Katalog # L11435.0B
Leverandør:   Thermo Fisher Scientific
Sorbitanmonostearat
Katalog # L11435.0B
CAS nummer:  1338-41-6
Sorbitanmonostearat
Sorbitanmonostearat

Glass bottle for solids, 1.000 g

CAS nummer:  1338-41-6
Price
1.010,00 kr
Log ind for tilgængelighed
1 * 1.000 g
Product Details & Documents

CAS No.: 1338-41-6 Molecular Formula: C24H46O6 Formula Weight: 430.63 Flash Point: 149°(300°F) Melting Point: 53-57° Merck Reference: 14,8724 MDL No.: MFCD00005366

Synonymer:  Sorbitan monooctadecanoate, (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol monooctadecanoate, Sorbitan stearate
  • CAS nummer:1338-41-6
  • Formel:C₂₄H₄₆O₆
  • MDL nummer:MFCD00005366
  • Molecular Weight:430.63 g/mol
  • Merck Index:14,8724
  • Smeltende Pt:53…57 °C
  • Tæthed:1,00
  • Flash Pt:>149 °C (300 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:215-664-9
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1.000 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.