Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ethylbenzen ≥99%

Ethylbenzen ≥99%

Glass bottle, 2.500 mL
Katalog # L05908.0F
Brandfarlig
Sundhed
Irriterende
Leverandør:   Thermo Fisher Scientific
Ethylbenzen ≥99%
Katalog # L05908.0F
CAS nummer:  100-41-4
Ethylbenzen ≥99%
Ethylbenzen ≥99%

Glass bottle, 2.500 mL

CAS nummer:  100-41-4
Price
992,00 kr
Log ind for tilgængelighed
1 * 2.500 mL
Product Details & Documents

Purity: 99% CAS No.: 100-41-4 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 15°(59°F) Refractive Index: 1.4950 Odour: Petrol-like Melting Point: -95° Boiling Point: 135-136° Beilstein Reference: 1901871 Merck Reference: 14,3765 UN No.: 1175 MDL No.: MFCD00011647

Synonymer:  Phenylethane
  • CAS nummer:100-41-4
  • Formel:C₈H₁₀
  • MDL nummer:MFCD00011647
  • Molecular Weight:106.17 g/mol
  • Merck Index:14,3765
  • Kogende Pt:135…136 °C
  • Smeltende Pt:–95 °C
  • Tæthed:0,867
  • Flash Pt:15 °C (59 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:202-849-4
  • FN:1175
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2.500 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.