Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cyclopentan for HPLC

Cyclopentan for HPLC

Glass bottle, 250 mL
Katalog # 44814.AK
Brandfarlig
Leverandør:   Thermo Fisher Scientific
Cyclopentan for HPLC
Katalog # 44814.AK
CAS nummer:  287-92-3
Cyclopentan for HPLC
Cyclopentan for HPLC

Glass bottle, 250 mL

CAS nummer:  287-92-3
Price
779,00 kr
Log ind for tilgængelighed
1 * 250 mL
Product Details & Documents

CAS No.: 287-92-3 Molecular Formula: C5H10 Formula Weight: 70.14 Flash Point: -37°(-34°F) Physical Form: Liquid Density: 0.748 Refractive Index: 1.4060 Appearance: Colourless Odour: Petrol-like Melting Point: -95° Boiling Point: 47-49° Beilstein Reference: 1900195 Merck Reference: 14,2741 UN No.: 1146 MDL No.: MFCD00001356

For HPLC and Spectrophotometry

Synonymer:  Pentamethylene
  • CAS nummer:287-92-3
  • Formel:C₅H₁₀
  • MDL nummer:MFCD00001356
  • Molecular Weight:70.13 g/mol
  • Merck Index:14,2741
  • Kogende Pt:47…49 °C
  • Smeltende Pt:–95 °C
  • Tæthed:0,748
  • Flash Pt:–37 °C (–34 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:206-016-6
  • FN:1146
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    250 mL

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.