Orders LinkContactUsLinkComponent
n-Hexadecan ≥99.5%
n-Hexadecan ≥99.5%
Glass bottle for solids, 5 g
Katalog # 44511.06
Sundhed
Irriterende
Leverandør:   Thermo Fisher Scientific
n-Hexadecan ≥99.5%
Katalog # 44511.06
CAS nummer:  544-76-3
n-Hexadecan ≥99.5%
n-Hexadecan ≥99.5%
Glass bottle for solids, 5 g
CAS nummer:  544-76-3
Price
1.120,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Purity: 99.5+% CAS No.: 544-76-3 Molecular Formula: CH3(CH2)14CH3 Formula Weight: 226.45 Flash Point: 135°(275°F) Physical Form: Liquid Density: 0.773 Refractive Index: 1.4345 Appearance: Colourless Melting Point: 18° Boiling Point: 287° Beilstein Reference: 1736592 MDL No.: MFCD00008998
Synonymer:  Hexadecane, Cetane
  • CAS nummer:544-76-3
  • Formel:H₃C(CH₂)₁₄CH₃
  • MDL nummer:MFCD00008998
  • Molecular Weight:226.45 g/mol
  • Kogende Pt:287 °C
  • Smeltende Pt:18 °C
  • Tæthed:0,773
  • Flash Pt:135 °C (275 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:208-878-9
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g
Specifikationer