Orders LinkContactUsLinkComponent
Stoppers, PTFE
Stoppers, PTFE
hvid, 29/32
Katalog # BOHLH937-07
Leverandør:   Bohlender
Stoppers, PTFE
Katalog # BOHLH937-07
Leverandør:  Bohlender
Stoppers, PTFE
Stoppers, PTFE
hvid, 29/32
Price
459,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
For closing ground joint parts. With ground joint and sealing rings on the outside; knurled or hexagonal grip. Compared with glass stoppers, they are easily removable and can be used without grease.
  • PTFE
  • Temperature resistance from −200 to + 250 °C
  • Chemical resistance
  • FDA conform
The stoppers can expand under heat which might lead to a breaking of the ground joint sockets.
Specifikationer
  • NS:
    29/32
  • Farve:
    hvid