Olie rød O
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 1320-06-5 Molecular Formula: C26H24N4O Formula Weight: 408.51 Melting Point: ca 166° dec. Beilstein Reference: 3491382 MDL No.: MFCD00003898
Synonyms: CI 26125
- CAS Number:1320-06-5
- Formula:C₂₆H₂₄N₄O
- MDL Number:MFCD00003898
- Molecular Weight:408.51 g/mol
- Melting Pt:∼ 166 °C (dec.)
- Storage Temperature:Stue temperatur
- EINECS:215-295-3
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:25 g



